Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-furoic Acid 98.0+%, TCI America™
SDP

Catalog No. B1008250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B1008250G 250 g
B100825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B1008250G Supplier TCI America Supplier No. B1008250G

Chemical Identifiers

CAS 585-70-6
Molecular Formula C5H3BrO3
Molecular Weight (g/mol) 190.98
MDL Number MFCD00003239
InChI Key YVTQHZDUDUCGRD-UHFFFAOYSA-N
Synonym 5-bromo-2-furoic acid, 5-bromofuroic acid, 5-bromo-furan-2-carboxylic acid, 5-bromo-2-furancarboxylic acid, 2-furancarboxylic acid, 5-bromo, 5-bromo-2-furanecarboxylic acid, 5-bromo furan-2-carboxylic acid, 2-furoic acid, 5-bromo, t5oj bvq ee, 5-bromofuroic?acid
PubChem CID 68511
IUPAC Name 5-bromofuran-2-carboxylic acid
SMILES C1=C(OC(=C1)Br)C(=O)O

Specifications

Melting Point 189°C
Color White-Yellow
Quantity 250 g
Formula Weight 190.98
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-furoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.