Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-hydroxy-3-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. B27971G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27971G 1 g
B27975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27971G Supplier TCI America Supplier No. B27971G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89488-30-2
Molecular Formula C6H6BrNO
Molecular Weight (g/mol) 188.024
InChI Key HXPMRPRBABWPKL-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxy-3-methylpyridine, 5-bromo-3-methylpyridin-2-ol, 5-bromo-2-hydroxy-3-picoline, 5-bromo-3-methylpyridin-2 1h-one, 5-bromo-3-methyl-pyridin-2-ol, 5-bromo-3-methyl-2-pyridinol, 2-hydroxy-5-bromo-3-picoline, 2-hydroxy-3-methyl-5-bromopyridine, 5-bromo-3-methyl-2-pyridone, 2 1h-pyridinone, 5-bromo-3-methyl
PubChem CID 285436
IUPAC Name 5-bromo-3-methyl-1H-pyridin-2-one
SMILES CC1=CC(=CNC1=O)Br

Specifications

Melting Point 172°C
Color Yellow
Quantity 1 g
Formula Weight 188.02
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-hydroxy-3-methylpyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.