Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-hydroxy-3-nitropyridine 98.0+%, TCI America™
SDP

Catalog No. B27061G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B27061G 1 g
B27065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B27061G Supplier TCI America Supplier No. B27061G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15862-34-7
Molecular Formula C5H3BrN2O3
Molecular Weight (g/mol) 218.994
InChI Key WXRLCVUDLFFTFF-UHFFFAOYSA-N
Synonym 5-bromo-2-hydroxy-3-nitropyridine, 5-bromo-3-nitropyridin-2-ol, 5-bromo-3-nitro-2-pyridinol, 5-bromo-3-nitro-2-pyridone, 5-bromo-3-nitropyridin-2 1h-one, 5-bromo-3-nitro-pyridin-2-ol, 3-bromo-6-hydroxy-5-nitropyridine, 5-bromo-3-nitro-2 1h-pyridinone, 5-bromo-3-nitro-2-hydroxypyridine, 2-hydroxy-3-nitro-5-bromopyridine
PubChem CID 85147
IUPAC Name 5-bromo-3-nitro-1H-pyridin-2-one
SMILES C1=C(C(=O)NC=C1Br)[N+](=O)[O-]

Specifications

Melting Point 247°C
Color Yellow
Quantity 1 g
Formula Weight 218.99
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-hydroxy-3-nitropyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.