Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-methoxy-4-methylpyridine 98.0+%, TCI America™
SDP

Catalog No. B40675G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B40675G 5 g
B40671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B40675G Supplier TCI America Supplier No. B40675G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 164513-39-7
Molecular Formula C7H8BrNO
Molecular Weight (g/mol) 202.051
MDL Number MFCD04039980
InChI Key HTBPXLJKMNBQMS-UHFFFAOYSA-N
PubChem CID 4178002
IUPAC Name 5-bromo-2-methoxy-4-methylpyridine
SMILES CC1=CC(=NC=C1Br)OC

Specifications

Melting Point 38°C
Color White-Yellow
Quantity 5 g
Formula Weight 202.05
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-methoxy-4-methylpyridine
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.