Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-2-thiophenesulfonamide 97.0+%, TCI America™
SDP

Catalog No. B279325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B279325G 25 g
B27935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B279325G Supplier TCI America Supplier No. B279325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53595-65-6
Molecular Formula C4H4BrNO2S2
Molecular Weight (g/mol) 242.11
MDL Number MFCD00067990
InChI Key WXJQQLDICAOBJB-UHFFFAOYSA-N
Synonym 5-bromo-2-thiophenesulfonamide, 5-bromo-thiophene-2-sulfonic acid amide, 2-thiophenesulfonamide, 5-bromo, 5-bromothiophene-2-sulfonic acid amide, 5-bromothiophene-2-sulphonamide, 2-bromo thiophene-5-sulfonamide, 5-bromothiophene-2-sulfonylamine, pubchem7369, maybridge1_004885, acmc-1b1sa
PubChem CID 1241304
IUPAC Name 5-bromothiophene-2-sulfonamide
SMILES NS(=O)(=O)C1=CC=C(Br)S1

Specifications

Melting Point 141°C
Color Yellow
Quantity 25 g
Formula Weight 242.11
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Bromo-2-thiophenesulfonamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.