Learn More
5-Bromo-4-chloro-3-indolyl Phosph., p-Toluidine Salt (Wht. to Off-Wht. Powd.), Fisher BioReagents
Shop All Fisherbrand ProductsDescription
Specifications
Specifications
| Alpha Vector | Bromo-4-chloro-3-indolyl Phosphate,p-Toluidine Salt (BCIP) |
| CAS | 1708766 |
| Melting Point | 222.8°C |
| Color | White |
| Assay Percent Range | ≥99 % |
| Packaging | Amber Glass |
| Quantity | 500 mg |
| Synonym | 4-nitrobenzo c 1,2,5 thiadiazole, 4-nitropiazthiole, 2,1,3-benzothiadiazole, 4-nitro, 4-nitro-1,2,3-benzothiadiazole, 4-nitro-benzo 1,2,5 thiadiazole, 4-nitro-2??,1,3-benzothiadiazole, 4-nitrobenzo c 1,2,5-thiadiazole, d0v8fg, acmc-1b59l, 4-bromo-2,8-dimethylphenol |
| Formula Weight | 433.62 |
| Percent Purity | 99% min. |
| Show More |
Safety and Handling
| Recommended Storage | <0°C |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.