Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-DL-tryptophan, 99%

Catalog No. AC107510050
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC107510050 5 g Glass Bottle
AC107515000 500 mg Glass Bottle
AC107510010 1 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC107510050 Supplier Thermo Scientific Chemicals Supplier No. 107510050
Add to Cart

CAS: 6548-09-0 | C11H11BrN2O2 | 283.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6548-09-0
Molecular Formula C11H11BrN2O2
Molecular Weight (g/mol) 283.13
MDL Number MFCD00005648
InChI Key KZDNJQUJBMDHJW-UHFFFAOYSA-N
Synonym 5-bromo-dl-tryptophan, 2-amino-3-5-bromo-1h-indol-3-yl propanoic acid, 5-bromotryptophan, tryptophan, 5-bromo, 5-bromo-tryptophan, 2-amino-3-5-bromoindol-3-yl propanoic acid, acmc-20a9ck, 2-amino-3-5-bromo-1h-indol-3-yl-propionic acid, 5-bromo-dl-tryptophan, sr-2-amino-3-5-bromoindolyl propionic acid
PubChem CID 96735
IUPAC Name 2-amino-3-(5-bromo-1H-indol-3-yl)propanoic acid
SMILES NC(CC1=CNC2=CC=C(Br)C=C12)C(O)=O

Specifications

Melting Point 261.0°C to 267.0°C
CAS Min % 98.5
CAS Max % 100.0
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Quantity 5 g
Formula Weight 283.12
Percent Purity 99%
Physical Form Fine Crystalline Powder
Chemical Name or Material 5-Bromo-DL-tryptophan
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.