Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromo-m-xylene 98.0+%, TCI America™
SDP

Catalog No. B179225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B179225G 25 g
B17925G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B179225G Supplier TCI America Supplier No. B179225G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 556-96-7
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000087
InChI Key LMFRTSBQRLSJHC-UHFFFAOYSA-N
Synonym 5-bromo-m-xylene, 3,5-dimethylbromobenzene, benzene, 1-bromo-3,5-dimethyl, m-xylene, 5-bromo, 5-bromo-1,3-xylene, 5-bromo-1,3-dimethylbenzene, 1-bromo-3,5-dimethyl-benzene, 5-bromo-meta-xylene, 5-bromo-3,5-dimethylbenzene, 1-bromo-3,5-dimethyl benzene
PubChem CID 136357
IUPAC Name 1-bromo-3,5-dimethylbenzene
SMILES CC1=CC(=CC(=C1)Br)C

Specifications

Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Boiling Point 205°C
Formula Weight 185.06
Percent Purity ≥98.0% (GC)
Chemical Name or Material 5-Bromo-m-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.