Learn More
Medchemexpress LLC 5-bromocytidine | 3066-86-2 | MFCD00057461 | 322.11 g·mol⁻1 | C9H12BrN3O5 | 5 G
SDP
Supplier:  Medchemexpress LLC HYW0222905G
Encompass_Preferred
5-Bromocytidine is a brominated cytidine nucleoside analogue used in biochemical and preclinical research to investigate DNA synthesis, nucleic acid metabolism, and nucleoside analogue activity. It is supplied in laboratory-scale pack sizes for experimental workflows and can be incorporated into assays studying nucleic acid modification and antiviral activity.
  • Brominated cytidine analogue for studies of DNA synthesis and modification.
  • Applicable to research in nucleic acid metabolism and antiviral assays.
  • Available in laboratory-scale pack sizes for flexible experimental design.
  • Molecular formula C9H12BrN3O5; molecular weight 322.11 g·mol⁻1.
  • Supplied with documentation sections for purity and handling information.
Catalog No. 50-003-00440