Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromopentyl Acetate 95.0+%, TCI America™
SDP

Catalog No. A1195100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A1195100G 100 g
A11955G 5 g
A119525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A1195100G Supplier TCI America Supplier No. A1195100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15848-22-3
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
MDL Number MFCD00039204
InChI Key JIHJLQWUVZUKCH-UHFFFAOYSA-N
Synonym 5-bromoamyl acetate, 5-bromo-n-amyl acetate, 1-acetoxy-5-bromopentane, 1-pentanol, 5-bromo-,1-acetate, 5-?bromopentyl acetate, acmc-209dhw, 5-bromopentylacetate, 5-bromo-1-pentanyl acetate, 5-bromopentyl acetate, acetic acid 5-bromoamyl ester
PubChem CID 85140
IUPAC Name 5-bromopentyl acetate
SMILES CC(=O)OCCCCCBr

Specifications

Color Yellow
Boiling Point 113°C
Quantity 100 g
Formula Weight 209.08
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Bromopentyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.