Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Bromophthalide, 98%

Catalog No. AAH3389114
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3389114 25 g
AAH3389106 5 g
AAH3389122 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAH3389114 Supplier Thermo Scientific Chemicals Supplier No. H3389114
Add to Cart

CAS: 64169-34-2 | C8H5BrO2 | 213.03 g/mol

5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Bromophthalide is an important organic intermediate. It can be used in agrochemical, pharmaceutical and dyestuff field. It is a key intermediate of citalopram.

Solubility
Soluble in acetone.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64169-34-2
Molecular Formula C8H5BrO2
Molecular Weight (g/mol) 213.03
MDL Number MFCD01797360
InChI Key IUSPXLCLQIZFHL-UHFFFAOYSA-N
Synonym 5-bromophthalide, 5-bromoisobenzofuran-1 3h-one, 5-bromo-1 3h-isobenzofuranone, 5-bromo-3h-isobenzofuran-1-one, 1 3h-isobenzofuranone, 5-bromo, 5-bromo-2-benzofuran-1 3h-one, 5-bromo-3-hydroisobenzofuran-1-one, 5-bromo-1,3-dihydro-2-benzofuran-1-one, 5-bromo-1,3-dihydro-isobenzofuran-1-one, 5-bromphthalide
PubChem CID 603144
IUPAC Name 5-bromo-1,3-dihydro-2-benzofuran-1-one
SMILES BrC1=CC=C2C(=O)OCC2=C1

Specifications

Melting Point 162°C to 166°C
Quantity 25 g
Solubility Information Soluble in acetone.
Formula Weight 213.03
Percent Purity 98%
Chemical Name or Material 5-Bromophthalide
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 447-190-5
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.