Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Carboxyfluorescein Diacetate 93.0+%, TCI America™
SDP

Catalog No. C2859200MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
50 mg
200 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C2859200MG 200 mg
C285950MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C2859200MG Supplier TCI America Supplier No. C2859200MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 79955-27-4
Molecular Formula C25H16O9
Molecular Weight (g/mol) 460.394
MDL Number MFCD00036872
InChI Key WPUZGNPQMIWOHE-UHFFFAOYSA-N
Synonym 5-carboxyfluorescein diacetate, 5-cfda, 3',6'-diacetoxy-3-oxo-3h-spiro isobenzofuran-1,9'-xanthene-5-carboxylic acid, 5-carboxy-di-o-acetylfluorescein, 3',6'-diacetyloxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, cfda-5, spiro isobenzofuran-1 3h ,9'-9h xanthene-5-carboxylicacid, 3',6'-bis acetyloxy-3-oxo, 3',6'-bis acetyloxy-3-oxospiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid, 3',6'-bis acetyloxy-3-oxospiro isobenzofuran-1 3h ,9'-9h xanthene-5-carboxylic acid, 3',6'-diacetoxy-3-oxo-3h-spiro 2-benzofuran-1,9'-xanthene-5-carboxylic acid
PubChem CID 133314
IUPAC Name 3',6'-diacetyloxy-3-oxospiro[2-benzofuran-1,9'-xanthene]-5-carboxylic acid
SMILES CC(=O)OC1=CC2=C(C=C1)C3(C4=C(C=C(C=C4)C(=O)O)C(=O)O3)C5=C(O2)C=C(C=C5)OC(=O)C

Specifications

Quantity 200 mg
Formula Weight 460.39
Percent Purity ≥93.0% (HPLC,T)
Chemical Name or Material 5-Carboxyfluorescein Diacetate
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.