Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 5-carboxyphthalide

Catalog No. CDS014604 Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CDS014604 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. CDS014604 Supplier Sigma Aldrich Supplier No. CDS014604100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Quantum chemical calculations of ground state energy, geometrical structure and vibrational wavenumbers of 1-acetylindole has been carried out using density functional (dft/b3lyp) method. regioselective acylations of 1-acetylindole (n-acetylindole) under friedel-crafts reaction has been reported. reaction of 1-acetylindole with manganese(iii) acetate in the presence of malonic acid, is reported to afford 4-acetyl-3,3a,4,8b-tetrahydro-2h-furo[3,2-b]indol-2-one.reactant for preparation of:
  • Antimycobacterial agents
  • Cyclin-dependent kinase (cdk2) inhibitorsreactant for:
  • C3-c3 oxidative cross-coupling reactionstandem mass spectrometry data independently generated by scripps center for metabolomics is available to view or download in pdf. 377104.pdf tested metabolites are featured on scripps center for metabolomics metlin metabolite database. to learn more, visit sigma.com/metlin

Specifications

CAS 4792-29-4
Molecular Formula C9H6O4
Linear Formula C9H6O4
MDL Number MFCD04117971
Quantity 5 g
Molecular Weight (g/mol) 178.14
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.