Learn More
AdipoGen 5-CFDA
SDP
Supplier:  AdipoGen CDXC0039M100
Encompass_Preferred
Chemical. CAS 79955-27-4. Formula C25H16O9. MW 460.39. Synthetic. Carboxyfluorescein diacetate CFDA, which was originally used to measure intracellular pH but was soon adapted for use as a cell viability indicator. Upon hydrolysis by intracellular nonspecific esterases, CFDA forms carboxyfluorescein. As compared with fluorescein, carboxyfluorescein contains extra negative charges and is therefore better retained in cells CFDA is moderately permeant to most cell membranes, with uptake greater at pH 6.2 than at pH 7.41. CFDA has been used as a viability probe with a variety of cells, including bacteria, fungi e.g. Saccharomyces cerevisiae, spermatozoa, natural killer NK cells and tumor cells.
Catalog No. 50-256-1527
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.