Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-1,3-dimethylpyrazole-4-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. C27471G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C27471G 1 g
C27475G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C27471G Supplier TCI America Supplier No. C27471G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 27006-76-4
Molecular Formula C6H7ClN2O
Molecular Weight (g/mol) 158.585
MDL Number MFCD00052538
InChI Key SZRSMNYUEXXEBL-UHFFFAOYSA-N
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole-4-carbaldehyde, 5-chloro-1,3-dimethyl-1h-pyrazole-4-carboxaldehyde, 5-chloro-1,3-dimethyl pyrazole-4-aldehyde, 5-chloro-1,3-dimethylpyrazole-4-carboxaldehyde, 5-chloro-4-formyl-1,3-dimethylpyrazole, 5-chloro-1,3-dimethyl-1h-pyrazole-4-aldehyde, 1h-pyrazole-4-carboxaldehyde, 5-chloro-1,3-dimethyl, pubchem23629, acmc-20a2nm, ksc496s9t
PubChem CID 2777397
IUPAC Name 5-chloro-1,3-dimethylpyrazole-4-carbaldehyde
SMILES CC1=NN(C(=C1C=O)Cl)C

Specifications

Melting Point 79°C
Color White-Yellow
Quantity 1 g
Formula Weight 158.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-1,3-dimethylpyrazole-4-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.