Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-1,3-dimethylpyrazole, 98+%, Thermo Scientific™

Catalog No. AC270220050
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC270220050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC270220050 Supplier Thermo Scientific Chemicals Supplier No. 270220050

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54454-10-3
Molecular Formula C5H7ClN2
Molecular Weight (g/mol) 130.58
MDL Number MFCD00051650
InChI Key DDUSLFAWARYAPR-UHFFFAOYSA-N
PubChem CID 521507
IUPAC Name 5-chloro-1,3-dimethyl-1H-pyrazole
SMILES CN1N=C(C)C=C1Cl

Specifications

CAS Min % 98.0
CAS Max % 100.0
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 98+%
Packaging Glass bottle
Refractive Index 1.4850 to 1.4870 (20°C, 589nm)
Quantity 5 g
Synonym 5-chloro-1,3-dimethyl-1h-pyrazole, 1h-pyrazole, 5-chloro-1,3-dimethyl, 1,3-dimethyl-5-chloropyrazole, pubchem9980, acmc-20am54, 5-chloro-1,3-dimethyl-pyrazole, 5-chloranyl-1,3-dimethyl-pyrazole, 1,3-dimethyl-5-chloro-1h-pyrazole, 1h-pyrazole,5-chloro-1,3-dimethyl, 5-chloro-1,3-dimethyl-1h-pyrazole #
Formula Weight 130.58
Percent Purity ≥98.0%
Physical Form Liquid
Chemical Name or Material 5-Chloro-1, 3-dimethylpyrazole, 98+%
Show More Show Less
Hazard Category Warning
Hazard Statement GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.