Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-1-pentanol 90.0+%, TCI America™
SDP

Catalog No. C081525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C081525ML 25 mL
C08155ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C081525ML Supplier TCI America Supplier No. C081525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 5259-98-3
Molecular Formula C5H11ClO
Molecular Weight (g/mol) 122.59
MDL Number MFCD00039556
InChI Key DCBJCKDOZLTTDW-UHFFFAOYSA-N
Synonym 5-chloro-1-pentanol, 5-chloropentanol, pentamethylene chlorohydrin, 1-pentanol, 5-chloro, 1-chloro-5-hydroxypentane, unii-55vp4w35eg, 1-chloro-5-pentanol, 5-chloropentane-1-ol, acmc-1amb4, ksc272o9l
PubChem CID 78915
IUPAC Name 5-chloropentan-1-ol
SMILES OCCCCCCl

Specifications

Color Yellow
Boiling Point 99°C
Quantity 25 mL
Formula Weight 122.59
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-1-pentanol
EINECSNumber (2)-2002
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.