Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-1-phenyl-1H-tetrazole 97%, Thermo Scientific™

Catalog No. AC162370500
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC162370500 50 g
AC162370100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC162370500 Supplier Thermo Scientific Chemicals Supplier No. 162370500

Item Discontinued This product is either new or inactive, and cannot be ordered online.

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14210-25-4
Molecular Formula C7H5ClN4
Molecular Weight (g/mol) 180.595
InChI Key DHELIGKVOGTMGF-UHFFFAOYSA-N
PubChem CID 84262
IUPAC Name 5-chloro-1-phenyltetrazole
SMILES C1=CC=C(C=C1)N2C(=NN=N2)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Quantity 50 g
Fieser 02,319; 04,377; 07,64; 08,102; 10,93
Synonym 5-chloro-1-phenyl-1h-tetrazole, 1h-tetrazole, 5-chloro-1-phenyl, 5-chloro-1-phenyl-1h-tetrazol, 1-phenyl-5-chlorotetrazole, 1h-tetrazole,5-chloro-1-phenyl, 5-chloro-1-phenyl-1h-1,2,3,4-tetrazole, 5-chloro-1-phenyl-1,2,3,4-tetraazole, acmc-209cnl, 1-phenyl-5-chlortetrazol, n-phenyl-5-chlorotetrazole
Formula Weight 180.6
Chemical Name or Material 5-Chloro-1-phenyl-1H-tetrazole, 97%
EINECSNumber 238-065-4
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.