Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2,4,6-trifluoropyrimidine 98.0+%, TCI America™
SDP

Catalog No. C166625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C166625G 25 g
C16665G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C166625G Supplier TCI America Supplier No. C166625G

Chemical Identifiers

CAS 697-83-6
Molecular Formula C4ClF3N2
Molecular Weight (g/mol) 168.50
MDL Number MFCD00191933
InChI Key GOYNRDSJTYLXBU-UHFFFAOYSA-N
Synonym pyrimidine, 5-chloro-2,4,6-trifluoro, 5-chloro-2,4,6-trifluoro-pyrimidine, 5-chlor-2,4,6-trifluorpyrimidin, 2,4,6-trifluoro-5-chloro pyrimidine, pubchem6908, acmc-1awug, 2,4,6-trifluoro-5-chloropyrimidine, 5-chloro-2,4-6-trifluoropyrimidine, 5-chloro-2,4,6-trifluoropyrimidine, 5-chloro-2,4,6-trifluoropyrimidine 5g
PubChem CID 69679
IUPAC Name 5-chloro-2,4,6-trifluoropyrimidine
SMILES FC1=NC(F)=C(Cl)C(F)=N1

Specifications

Color Yellow
Boiling Point 115°C
Quantity 25 g
Formula Weight 168.50
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 5-Chloro-2,4,6-trifluoropyrimidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.