Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Chloro-2'-hydroxy-4'-methylacetophenone 95.0+%, TCI America™
SDP

Catalog No. C162925G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C162925G 25 g
C16295G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C162925G Supplier TCI America Supplier No. C162925G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 28480-70-8
Molecular Formula C9H9ClO2
Molecular Weight (g/mol) 184.619
MDL Number MFCD00191920
InChI Key HDUSGGZSLVCDKY-UHFFFAOYSA-N
Synonym 4-Acetyl-2-chloro-5-hydroxytoluene, 2-Acetyl-4-chloro-5-methylphenol
PubChem CID 598229
IUPAC Name 1-(5-chloro-2-hydroxy-4-methylphenyl)ethanone
SMILES CC1=C(C=C(C(=C1)O)C(=O)C)Cl

Specifications

Melting Point 69°C
Color Yellow
Boiling Point 137°C
Quantity 25 g
Formula Weight 184.62
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Chloro-2′-hydroxy-4′-methylacetophenone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.