Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-hydroxypyridine 98.0+%, TCI America™
SDP

Catalog No. C124525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C124525G 25 g
C12455G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C124525G Supplier TCI America Supplier No. C124525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 4214-79-3
Molecular Formula C5H4ClNO
Molecular Weight (g/mol) 129.54
MDL Number MFCD00006275
InChI Key SZFUWUOHDRMCKD-UHFFFAOYSA-N
Synonym 5-chloro-2-hydroxypyridine, 5-chloropyridin-2-ol, 5-chloro-2-pyridinol, 5-chloropyridin-2 1h-one, 2-hydroxy-5-chloropyridine, 5-chloro-2-pyridol, 5-chloro-2 1h-pyridinone, 5-chloro-2-pyridone, 2 1h-pyridinone, 5-chloro, unii-r7a6f5d3ky
PubChem CID 77889
IUPAC Name 5-chloro-1,2-dihydropyridin-2-one
SMILES ClC1=CNC(=O)C=C1

Specifications

Melting Point 164°C
Color White-Yellow
Quantity 25 g
Formula Weight 129.54
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-hydroxypyridine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.