Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methoxybenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C1539500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1539500G 500 g
C153925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1539500G Supplier TCI America Supplier No. C1539500G

Chemical Identifiers

CAS 3438-16-2
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.591
MDL Number MFCD00017546
InChI Key HULDRQRKKXRXBI-UHFFFAOYSA-N
Synonym 5-chloro-o-anisic acid, 5-chloro-2-methoxy-benzoic acid, benzoic acid, 5-chloro-2-methoxy, 5-chloro-2-methoxybenzoicacid, 2-methoxy-5-chlorobenzoic acid, 5-chloro-2-anisic acid, pubchem2680, 2-carboxy-4-chloroanisole, 5-chloro-2-methoxy-benzoate, 5-chloro-methoxybenzoic acid
PubChem CID 76970
IUPAC Name 5-chloro-2-methoxybenzoic acid
SMILES COC1=C(C=C(C=C1)Cl)C(=O)O

Specifications

Melting Point 99°C
Quantity 500 g
Formula Weight 186.59
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methoxybenzoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.