Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methylaniline 98.0+%, TCI America™
SDP

Catalog No. C1876500G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1876500G 500 g
C187625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C1876500G Supplier TCI America Supplier No. C1876500G

Chemical Identifiers

CAS 95-79-4
Molecular Formula C7H8ClN
Molecular Weight (g/mol) 141.60
MDL Number MFCD00007779
InChI Key WRZOMWDJOLIVQP-UHFFFAOYSA-N
Synonym 5-chloro-o-toluidine, 2-amino-4-chlorotoluene, 2-methyl-5-chloroaniline, fast red kb base, benzenamine, 5-chloro-2-methyl, ansibase red kb, red kb base, 4-chloro-2-aminotoluene, 3-chloro-6-methylaniline, pharmazoid red kb
PubChem CID 7260
ChEBI CHEBI:82422
IUPAC Name 5-chloro-2-methylaniline
SMILES CC1=CC=C(Cl)C=C1N

Specifications

Color White-Yellow
Boiling Point 237°C
Quantity 500 g
UN Number 2239
Formula Weight 141.60
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Lumps
Chemical Name or Material 5-Chloro-2-methylaniline
EINECSNumber (3)-0265
RTECSNumber XU5075000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.