Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-methylbenzoxazole 98.0+%, TCI America™
SDP

Catalog No. C1276100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C1276100G 100 g
C127625G 25 g
C1276500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C1276100G Supplier TCI America Supplier No. C1276100G

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

Chemical Identifiers

CAS 19219-99-9
Molecular Formula C8H6ClNO
Molecular Weight (g/mol) 167.592
MDL Number MFCD00022850
InChI Key HJCIGAUHTJBHBQ-UHFFFAOYSA-N
PubChem CID 29510
IUPAC Name 5-chloro-2-methyl-1,3-benzoxazole
SMILES CC1=NC2=C(O1)C=CC(=C2)Cl

Specifications

Melting Point 57°C
Boiling Point 220°C
Quantity 100 g
Formula Weight 167.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-methylbenzoxazole
RTECSNumber DM4790000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.