Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-chloro-2-(methylthio)aniline, 95%, Thermo Scientific™

Catalog No. CD05201DA
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CD05201DA 1 g Amber Glass Bottle
CD05201EA 10 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CD05201DA Supplier Thermo Scientific Chemicals Supplier No. CD05201DA
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 16423-54-4
Molecular Formula C7H8ClNS
Molecular Weight (g/mol) 173.658
MDL Number MFCD00052816
InChI Key RWUQNIPYCAHKMO-UHFFFAOYSA-N
PubChem CID 2794577
IUPAC Name 5-chloro-2-methylsulfanylaniline
SMILES CSC1=C(C=C(C=C1)Cl)N

Specifications

Assay Percent Range 95%
Packaging Amber Glass Bottle
Linear Formula NH2C6H3(Cl)SCH3
Quantity 1 g
Synonym 5-chloro-2-methylthio aniline, 5-chloro-2-methylsulfanyl aniline, 5-chloro-2-methylthiophenylamine, 5-chloro-2-methylthio benzenamine, 5-chloro-2-methylsulfanyl-phenylamine, benzenamine,5-chloro-2-methylthio, benzenamine, 5-chloro-2-methylthio
Formula Weight 173.67
Percent Purity 95%
Chemical Name or Material 5-Chloro-2-(methylthio)aniline
Hazard Statement GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Harmful if inhaled.
May cause respiratory irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.