Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-nitrobenzoic Acid 98.0+%, TCI America™
SDP

Catalog No. C0224250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0224250G 250 g
C022425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0224250G Supplier TCI America Supplier No. C0224250G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2516-95-2
Molecular Formula C7H4ClNO4
Molecular Weight (g/mol) 201.562
MDL Number MFCD00007290
InChI Key ZKUYSJHXBFFGPU-UHFFFAOYSA-N
Synonym 5-chloro-2-nitrobenzoic acid, benzoic acid, 5-chloro-2-nitro, 2-nitro-5-chlorobenzoic acid, 3-chloro-6-nitrobenzoic acid, 5-chloro-2-nitro-benzoic acid, pubchem4586, acmc-209ghs, rarechem al bo 1106, timtec-bb sbb009922, 2-nitro-5-chlorobenzoate
PubChem CID 17286
SMILES C1=CC(=C(C=C1Cl)C(=O)O)[N+](=O)[O-]

Specifications

Melting Point 139°C
Color Yellow
Quantity 250 g
Formula Weight 201.56
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitrobenzoic Acid
EINECSNumber (3)-1502
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.