Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-2-nitrodiphenylamine 98.0+%, TCI America™
SDP

Catalog No. C178525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C178525G 25 g
C17855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C178525G Supplier TCI America Supplier No. C178525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 25781-92-4
Molecular Formula C12H9ClN2O2
Molecular Weight (g/mol) 248.666
MDL Number MFCD00007287
InChI Key FPKHZBVGKMTUHB-UHFFFAOYSA-N
PubChem CID 117635
IUPAC Name 5-chloro-2-nitro-N-phenylaniline
SMILES C1=CC=C(C=C1)NC2=C(C=CC(=C2)Cl)[N+](=O)[O-]

Specifications

Melting Point 114°C
Quantity 25 g
Synonym 5-chloro-2-nitrodiphenylamine, benzenamine, 5-chloro-2-nitro-n-phenyl, 5-chloro-2-nitrophenyl phenylamine, ghl.pd_mitscher_leg0.920, ksc203m3b, 2-anilino-4-chloronitrobenzene, fpkhzbvgkmtuhb-uhfffaoysa, 5-chloro-2-nitro-n-phenyl-benzenamine, benzenamine,5-chloro-2-nitro-n-phenyl
Formula Weight 248.67
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloro-2-nitrodiphenylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.