Learn More
Sigma Aldrich 5-Chloro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1-(triisopropylsilyl)-1H-pyrrolo[2,3-b]pyridine
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
- Methyl 2,6-difluorobenzoate is a fluorinated aromatic ester
- It can be synthesized from 2,6-difluorobenzoic acid via esterification with methanol.methyl 2,6-difluorobenzoate may be used in the synthesis of 9-hydroxy-13-(2,6-difluorophenyl)-3h-dibenzo[b,i]xanthen-3-one and 2,6-difluorobenzohydroxamic acid
Specifications
Specifications
| CAS | 1072152-34-1 |
| Molecular Formula | C22H36BClN2O2Si |
| Linear Formula | C22H36BClN2O2Si |
| MDL Number | MFCD09965900 |
| Quantity | 25 g |
| Molecular Weight (g/mol) | 434.88 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.