Learn More
Medchemexpress LLC 5-chloro-7-[(3-chlorophenyl)(2-pyridinylamino)methyl]-8-quinolinol | 709009-15-4 | 98.0% | 396.27 | C21H15Cl2N3O | 25MG

Supplier: Medchemexpress LLC HY12479125MG
MMRi6 (5-Chloro-7-[(3-chlorophenyl)(2-pyridinylamino)methyl]-8-quinolinol) is a small-molecule research inhibitor that disrupts Mdm2-MdmX RING-RING interactions and modulates the p53 pathway. It is supplied for laboratory research use only. Typical analytical purity is 98% (HPLC). Molecular formula C21H15Cl2N3O; molecular weight 396.27 g/mol. Recommended storage: 4°C, sealed, away from moisture and light; in solvent: -80°C (6 months) or -20°C (1 month).
- Disrupts Mdm2-MdmX RING-RING interaction in vitro.
- Suitable for p53 pathway and ubiquitination studies.
- Analytical purity 98% by HPLC.
- Molecular formula C21H15Cl2N3O; molecular weight 396.27 g/mol.
- Recommended storage: 4°C sealed; in solvent -80°C (6 months) or -20°C (1 month).
- Available in small research quantities (for example, 1 mg, 5 mg, 10 mg).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.