Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloro-7-methylisatin, 97%

Catalog No. AAL1168206
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1168206 5 g
AAL11682MD 250 mg
AAL1168203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1168206 Supplier Thermo Scientific Chemicals Supplier No. L1168206
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 14389-06-1 | C9H6ClNO2 | 195.602 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14389-06-1
Molecular Formula C9H6ClNO2
Molecular Weight (g/mol) 195.602
MDL Number MFCD00047218
InChI Key LDFQLYHDZZPAGN-UHFFFAOYSA-N
Synonym 5-chloro-7-methylisatin, 5-chloro-7-methylindoline-2,3-dione, 5-chloro-7-methyl-2,3-dihydro-1h-indole-2,3-dione, 5-chloro-7-methyl-1h-benzo d azolidine-2,3-dione, pubchem13601, acmc-1buvx, isatin-based compound, 56, 5-chloro-7-methyl isatin, 1h-indole-2,3-dione,5-chloro-7-methyl, 5-chloranyl-7-methyl-1h-indole-2,3-dione
PubChem CID 366710
IUPAC Name 5-chloro-7-methyl-1H-indole-2,3-dione
SMILES CC1=C2C(=CC(=C1)Cl)C(=O)C(=O)N2

Specifications

Melting Point ∽283°C (decomposition)
Quantity 5 g
Beilstein 153061
Formula Weight 195.61
Percent Purity 97%
Chemical Name or Material 5-Chloro-7-methylisatin
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.