Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chlorobenzotriazole 98.0+%, TCI America™
SDP

Catalog No. C0137100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C0137100G 100 g
C013725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C0137100G Supplier TCI America Supplier No. C0137100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 94-97-3
Molecular Formula C6H4ClN3
Molecular Weight (g/mol) 153.57
MDL Number MFCD00005700
InChI Key PZBQVZFITSVHAW-UHFFFAOYSA-N
Synonym 5-chlorobenzotriazole, 1h-benzotriazole, 5-chloro, 6-chlorobenzotriazole, 5-chloro-1h-benzotriazole, 1h-benzotriazole, 6-chloro, 5-chloro-1h-benzo d 1,2,3 triazole, 5-chloro-1h-1,2,3-benzotriazole, 6-chloro-1h-benzotriazole, unii-8c7o46g78k, 5-chlorobenztriazole
PubChem CID 66760
IUPAC Name 5-chloro-2H-1,2,3-benzotriazole
SMILES ClC1=CC2=NNN=C2C=C1

Specifications

Melting Point 158°C
Color White-Yellow
Quantity 100 g
Formula Weight 153.57
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chlorobenzotriazole
EINECSNumber (5)-0540
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.