Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloroindole 98.0+%, TCI America™
SDP

Catalog No. C110725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C110725G 25 g
C11075G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C110725G Supplier TCI America Supplier No. C110725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 17422-32-1
Molecular Formula C8H6ClN
Molecular Weight (g/mol) 151.593
MDL Number MFCD00005672
InChI Key MYTGFBZJLDLWQG-UHFFFAOYSA-N
Synonym 5-chloroindole, 1h-indole, 5-chloro, 5-chloro indole, 1h-indole, 5-chloro-9ci, 5-chloroindol, 5-chloro-indole, 3fue, zlchem 215, 5-chlor-1h-indole, indole, 5-chloro
PubChem CID 87110
IUPAC Name 5-chloro-1H-indole
SMILES C1=CC2=C(C=CN2)C=C1Cl

Specifications

Melting Point 72°C
Boiling Point 130°C
Quantity 25 g
Formula Weight 151.59
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Chloroindole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.