Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloroisatin, 98%

Catalog No. AAA1312514
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1312514 25 g
AAA1312506 5 g
AAA1312522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1312514 Supplier Thermo Scientific Chemicals Supplier No. A1312514
Add to Cart

CAS: 17630-76-1 | C8H4ClNO2 | 181.58 g/mol

5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloroisatin is a reagent used in the synthesis of Schiff bases through the reaction with 2-methyl-4-nitroaniline or 4-amino-4,5-dihydro-1H-1,2,3-triazole-5-one.

Solubility
It is insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17630-76-1
Molecular Formula C8H4ClNO2
Molecular Weight (g/mol) 181.58
MDL Number MFCD00014567
InChI Key XHDJYQWGFIBCEP-UHFFFAOYSA-N
Synonym 5-chloroisatin, 5-chloroindoline-2,3-dione, 5-chlorisatin, 1h-indole-2,3-dione, 5-chloro, 5-chloro-2,3-dihydro-1h-indole-2,3-dione, 5-chloro isatin, 5-chlor-2,3-dioxoindolin, 5-chlor-1h-indol-2,3-dion, 2hq, pubchem13607
PubChem CID 87203
SMILES ClC1=CC=C2NC(=O)C(=O)C2=C1

Specifications

Melting Point ∼247°C (decomposition)
Quantity 25 g
Beilstein 383759
Solubility Information It is insoluble in water.
IUPAC Name 5-chloro-1H-indole-2,3-dione
Formula Weight 181.58
Percent Purity 98%
Chemical Name or Material 5-Chloroisatin
EINECSNumber 241-614-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.