Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Chloromethyl-2-(1H-pyrazol-1-yl)pyridine, 97%, Thermo Scientific Chemicals

Catalog No. AAH6179206
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6179206 5 g
AAH6179203 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6179206 Supplier Thermo Scientific Chemicals Supplier No. H6179206

5-Chloromethyl-2-(1H-pyrazol-1-yl)pyridine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
5-Chloromethyl-2-(1H-pyrazol-1-yl)pyridine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Light sensitive. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 748796-39-6
Molecular Formula C9H8ClN3
Molecular Weight (g/mol) 193.634
MDL Number MFCD06658399
InChI Key QVDLCABKNWXXQF-UHFFFAOYSA-N
PubChem CID 16655946
IUPAC Name 5-(chloromethyl)-2-pyrazol-1-ylpyridine
SMILES C1=CN(N=C1)C2=NC=C(C=C2)CCl

Specifications

Color White
Quantity 5 g
Sensitivity Light sensitive
Synonym 5-chloromethyl-2-1h-pyrazol-1-yl pyridine, 5-chloromethyl-2-pyrazol-1-yl pyridine, 5-chloromethyl-2-pyrazol-1-yl-pyridine, 3,4-difluorophenyl methyl methyl amine, pyridine,5-chloromethyl-2-1h-pyrazol-1-yl, pyridine, 5-chloromethyl-2-1h-pyrazol-1-yl
Solubility Information Slightly soluble in water.
Formula Weight 193.63
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 5-Chloromethyl-2-(1H-pyrazol-1-yl)pyridine
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
storageNote1 2° to 8°C
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.