Learn More
Sigma Organic Chemistry 5- Chloromethyl -3-isop | 1G | MFCD08691639
Supplier: Sigma Organic Chemistry CBR019681G

5- Chloromethyl -3-isop , 1G
About this Item:
MDL #: MFCD08691639
Molecular Weight: 153.22
UNSPSC Code: 12352200
Chemical Formula: C8H15N3
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.