Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyanoindole 98.0+%, TCI America™
SDP

Catalog No. C149125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C149125G 25 g
C14911G 1 g
C14915G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. C149125G Supplier TCI America Supplier No. C149125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 15861-24-2
Molecular Formula C9H6N2
Molecular Weight (g/mol) 142.161
MDL Number MFCD00005669
InChI Key YHYLDEVWYOFIJK-UHFFFAOYSA-N
Synonym 5-cyanoindole, indole-5-carbonitrile, 5-cyano indole, 5-cyano-1h-indole, indole-5-cyano, 5-indolecarbonitrile, 5-cyano-indole, indol-5-carbonitrile, indole, 5-cyano, pubchem7326
PubChem CID 27513
IUPAC Name 1H-indole-5-carbonitrile
SMILES C1=CC2=C(C=CN2)C=C1C#N

Specifications

Melting Point 106°C
Quantity 25 g
Formula Weight 142.16
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 5-Cyanoindole
RTECSNumber NL5993120
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.