Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyanophthalide 98.0+%, TCI America™
SDP

Catalog No. C200125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C200125G 25 g
C20015G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C200125G Supplier TCI America Supplier No. C200125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 82104-74-3
Molecular Formula C9H5NO2
Molecular Weight (g/mol) 159.144
InChI Key XEEGWTLAFIZLSF-UHFFFAOYSA-N
Synonym 5-cyanophthalide, 1-oxo-1,3-dihydroisobenzofuran-5-carbonitrile, 5-cyano phthalide, 5-cyano-3h-isobenzofuranone, 1,3-dihydro-1-oxoisobenzofuran-5-carbonitrile, 1-oxo-1,3-dihydro-2-benzofuran-5-carbonitrile, 1-oxo-3h-isobenzofuran-5-carbonitrile, 1,3-dihydro-1-oxo-5-isobenzofurancarbonitrile, 5-isobenzofurancarbonitrile, 1,3-dihydro-1-oxo, 5-cyanophthalid
PubChem CID 821218
IUPAC Name 1-oxo-3H-2-benzofuran-5-carbonitrile
SMILES C1C2=C(C=CC(=C2)C#N)C(=O)O1

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 25 g
Formula Weight 159.14
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material 5-Cyanophthalide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.