Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Cyclobutyl-5-phenylhydantoin, 97+%, Thermo Scientific™

Catalog No. AAA1611503
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1611503 1 g
AAA1611506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1611503 Supplier Thermo Scientific Chemicals Supplier No. A1611503

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 125650-44-4
Molecular Formula C13H14N2O2
Molecular Weight (g/mol) 230.27
MDL Number MFCD00022395
InChI Key YFMSPNSDLBGLRF-UHFFFAOYNA-N
Synonym 5-cyclobutyl-5-phenylhydantoin, acmc-20am3v, 5-cyclobutyl-5-phenyl-1,3-diazolidine-2,4-dione
PubChem CID 277170
IUPAC Name 5-cyclobutyl-5-phenylimidazolidine-2,4-dione
SMILES O=C1NC(=O)C(N1)(C1CCC1)C1=CC=CC=C1

Specifications

Melting Point 234°C to 235°C
Quantity 1 g
Formula Weight 230.27
Percent Purity ≥97%
Chemical Name or Material 5-Cyclobutyl-5-phenylhydantoin
TSCA No
Recommended Storage Ambient temperatures
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.