Learn More
Sigma Organic Chemistry 5-Cyclohexyl-1 3 4-thia | 1G | 56882-77-0 | MFCD00469715
Supplier: Sigma Organic Chemistry TMT002851G

5-Cyclohexyl-1 3 4-thia , 1G
About this item:
CAS #: 56882-77-0
MDL #: MFCD00469715
Chemical Formula: C8H13N3S
Molecular Weight: 183.27
UNSPSC Code: 12352200
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.