Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5'-Deoxy-5-fluorocytidine 98.0+%, TCI America™
SDP

Catalog No. D43421G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43421G 1 g
D43425G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43421G Supplier TCI America Supplier No. D43421G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 66335-38-4
Molecular Formula C9H12FN3O4
Molecular Weight (g/mol) 245.21
MDL Number MFCD07369278
InChI Key YSNABXSEHNLERR-ZIYNGMLESA-N
PubChem CID 10037499
ChEBI CHEBI:80627
IUPAC Name 4-amino-1-[(2R,3R,4S,5R)-3,4-dihydroxy-5-methyloxolan-2-yl]-5-fluoro-1,2-dihydropyrimidin-2-one
SMILES C[C@H]1O[C@H]([C@H](O)[C@@H]1O)N1C=C(F)C(N)=NC1=O

Specifications

Melting Point 204°C
Color White
Quantity 1 g
Formula Weight 245.21
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5′-Deoxy-5-fluorocytidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.