Learn More
Sigma Organic Chemistry 5-Diisopropylamino amyl | 100mg | MFCD00048285
Supplier: Sigma Organic Chemistry CDS000708100MG

5-Diisopropylamino amyl , 100mg
About this Item:
MDL #: MFCD00048285
Molecular Weight: 186.34
UNSPSC Code: 12352200
Chemical Formula: C11H26N2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.