Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

5-Dimethylamino-2-nitrosophenol Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. N026125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N026125G 25 g
N02615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N026125G Supplier TCI America Supplier No. N026125G

Chemical Identifiers

CAS 41317-10-6
Molecular Formula C8H11ClN2O2
Molecular Weight (g/mol) 202.638
MDL Number MFCD00060162
InChI Key WRIZNEWVHJJMHU-UHFFFAOYSA-N
Synonym 2-Nitroso-5-dimethylaminophenol Hydrochloride
PubChem CID 9815614
IUPAC Name 5-(dimethylamino)-2-nitrosophenol;hydrochloride
SMILES CN(C)C1=CC(=C(C=C1)N=O)O.Cl

Specifications

Quantity 25 g
Formula Weight 202.64
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 5-Dimethylamino-2-nitrosophenol Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.