Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals 5'-Ethynyl-2'-deoxyuridine

252.23 g/mol

$212.50 - $543.65

Specifications

Molecular Weight (g/mol) 252.23
Product Type 5 '-Ethynyl-2 '-deoxyuridine
Solubility Information Solubility aq. soln.: .. Other solubilities: soluble in dmso, methanol

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
AC466380010
SDS View Documents
Thermo Scientific Chemicals
466380010
1 g
Each for $543.65

Save 23%

Reg : $708.50

The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Order in Bulk

Request bulk or custom formats

Only null left
Add to Cart
 
AC466382500
SDS View Documents
Thermo Scientific Chemicals
466382500
250 mg
Each for $212.50
Order in Bulk

Request bulk or custom formats

Only null left
Add to Cart
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

252.23
5 '-Ethynyl-2 '-deoxyuridine
Solubility aq. soln.: .. Other solubilities: soluble in dmso, methanol
Videos
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.