Learn More
Abcam 5-Ethynyl-dUTP, Aqueous Solution, >95% Purity, 25 uL
Description
- Molecular Weight: 492.16 g/mol
- Purity: >95%
- Application: Incorporated into DNA by Primer Extension with family A polymerase Taq and family B polymerases
- Format: Aqueous solution
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Substrate Properties | Chemical Substrate |
| Target Molecule | DNA |
| Label or Dye | Unconjugated |
| Format | Tube |
| Buffer | aqueous solution |
| Form | Solution |
| Formulation | aqueous solution |
| Quantity | 25 μL |
| For Use With (Application) | Primer Extension |
| Storage Requirements | Store at -20°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.