Learn More
Abcam 5-Ethynyl-UTP (5-EUTP) (aqueous solution), Nucleotide analog, 5UL
Supplier:  Abcam AB1467445UL
Encompass_Preferred
MW 507.97 g/mol, Purity >95%. Nucleotide analog. Incorporated into RNA by T7 RNA polymerase-mediated in vitro transcription to produce alkyne-functionalized RNA.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-3219