Learn More
TARGETMOL CHEMICALS INC 5-Feruloylquinic acid 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN12961MG
Encompass_Preferred
Also available in 5 mg, 10 mg, 25 mg, 50 mg and bulk. Please contact Fisher for quotes. 5-Feruloylquinic acid (5-FQA) is a potent Sirt1 agonist and a potential lead compound for further testing in the drug development process for aging-associated diseases. Purity 99.11%
Catalog No. 50-287-9771
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.