Learn More
TARGETMOL CHEMICALS INC Azaleatin 1MG
SDP
Supplier:  TARGETMOL CHEMICALS INC TN14181MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Azaleatin is a natural product from Rhododendron simsii. Azaleatin is a dipeptidyl peptidase-IV inhibitor. Azaleatin can be used for type-2 diabetes and obesity studies. Purity 98.62%
Catalog No. 50-287-9772
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.