Learn More
Abcam 5-FITC (5-Fluorescein Isothiocyanate), Fluorescent marker, 250MG
Supplier:  Abcam AB145325250MG
Encompass_Preferred
MW 389.4 Da, Purity >98%. Fluorescent marker. Amine reactive fluorescein isomer. Yields conjugates with superior stability compared to other FITC isomers. Donor for FRET assays.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-5287
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.