Learn More
AdipoGen 5-Fluoresceinyl-maleinimide
SDP
Supplier:  AdipoGen CDXF0004M500
Encompass_Preferred
Chemical. CAS 75350-46-8. Formula C24H13NO7. MW 427.36. Synthetic. Green fluorescent thiol-reactive dye Ex/Em 494/519nm for bioconjugation, widely used as protein and peptide fluorescent labeling reagent, easily conjugating fluorescein. The reaction requires very mild condition and is highly specific. The maleimide group reacts optimally with sulfhydryl groups on cysteine side chains at pH 7, forming a stable thioether bond. The compound can be used to label proteins for the detection of conformational changes, assembly of multisubunit complexes, and ligand-binding processes.
Catalog No. 50-256-0949
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.