Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ 5' Fluorescent Labeled Oligo

Catalog No. 450006H
Encompass_Preferred
Change view
Click to view available options
Product Type:
5' Labeled Primers
Quantity:
80,000 pmol
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type Quantity
45-000-6H 5' Labeled Primers 80,000 pmol
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-6H Supplier Invitrogen™ Supplier No. 450006H
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

5' Fluorescent Labeled Oligo

Custom 5'-labeled primers are ideal for applications including fragment analysis, gene expression studies, genotyping, and PCR. Choose among FAM™, TET™, VIC™, HEX™, NED™, and PET™ as fluorescent 5' labels. Oligos are desalted; HPLC purification is also available.
TRUSTED_SUSTAINABILITY

Specifications

Delivery Type Solution or Dried-down
Description 5' Labeled Primers, 1 Tube, 40,000 pmol HPLC Yield, 6-FAM™, TET™, VIC™, HEX™, NED™, PET™ 5' Reporter Dye, 12 Months Shelf Life, Room Temperature Shipping Conditions, –20°C Storage Temperature, For Fragment Analysis, Gene Expression Studies, Genotyping and PCR
Product Type 5' Labeled Primers
Length (Metric) 1.524 m
Content And Storage Store in Freezer (-5°C to -30°C).
Purification Method Desalted, Option of HPLC Purification with Larger Scale
Label or Dye FAM, HEX, NED, PET, TET, VIC
Primer 5' Labeled
Quantity 80,000 pmol
Grade HPLC
For Use With (Application) Fragment Analysis, Gene Expression Studies, Genotyping, PCR
Form Delivered either in Solution or Dried down
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.